Ethyl-4-chloro-1-methyl-6-oxo-1,6-dihydro-3-pyridinecarboxylate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-chloro-1-methyl-6-oxo-1,6-dihydropyridine-3-carboxylate

A169267-10G A169267-1G A169267-250MG
77286-742EA 682.89 USD
77286-742 77286-744 77286-746
Ethyl-4-chloro-1-methyl-6-oxo-1,6-dihydro-3-pyridinecarboxylate 97%
Ethyl-4-chloro-1-methyl-6-oxo-1,6-dihydro-3-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₃
MW: 215.64 g/mol
MDL Number: MFCD08234670
CAS Number: 821791-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR