Diacetin 40%

Supplier: Ambeed
Warning

A169239-100G A169239-500G
77247-932EA 8 USD
77247-932 77247-934
Diacetin 40%
Diacetin

Precaución: for Research Use Only

Formula: C₇H₁₂O₅
MDL Number: MFCD00008717
CAS Number: 25395-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR