3-Bromo-7-methylimidazo[1,2-a]pyrimidine 98%

Supplier: Ambeed
Warning

A169163-100MG A169163-1G A169163-250MG
77273-640EA 50.47 USD
77273-640 77273-642 77273-644
3-Bromo-7-methylimidazo[1,2-a]pyrimidine 98%
3-Bromo-7-methylimidazo[1,2-a]pyrimidine

Precaución: for Research Use Only

Formula: C7H6BrN3
MW: 212.0500 g/mol
MDL Number: MFCD08234998
CAS Number: 375857-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR