5-Amino-7-(cyclohexylamino)-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 98%

Supplier: Ambeed
Danger

A169136-10MG A169136-1MG A169136-250MG A169136-50MG A169136-5MG
77259-866EA 113.3 USD
77259-866 77259-868 77259-870 77259-872 77259-874
5-Amino-7-(cyclohexylamino)-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid 98%
5-Amino-7-(cyclohexylamino)-1-ethyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₂FN₃O₃
MW: 347.38 g/mol
MDL Number: MFCD30718173
CAS Number: 836620-48-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR