tert-Butyl 4-(3-iodobenzoyl)piperazine-1-carboxylate 98%

Supplier: Ambeed
Warning

A169008-10G A169008-1G A169008-250MG A169008-25G A169008-5G
77236-196EA 363 USD
77236-196 77236-198 77236-200 77236-202 77236-204
tert-Butyl 4-(3-iodobenzoyl)piperazine-1-carboxylate 98%
tert-Butyl 4-(3-iodobenzoyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₆H₂₁IN₂O₃
MW: 416.25 g/mol
MDL Number: MFCD14635742
CAS Number: 838845-52-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR