5-Fluoro-4,7-bis(5-(trimethylstannyl)thiophen-2-yl)benzo[c][1,2,5]thiadiazole 97%

Supplier: Ambeed
Danger

A168997-100MG A168997-250MG
77247-920EA 49.44 USD
77247-920 77247-922
5-Fluoro-4,7-bis(5-(trimethylstannyl)thiophen-2-yl)benzo[c][1,2,5]thiadiazole 97%
5-Fluoro-4,7-bis(5-(trimethylstannyl)thiophen-2-yl)benzo[c][1,2,5]thiadiazole

Precaución: for Research Use Only

Formula: C₂₀H₂₃FN₂S₃Sn₂
MW: 644.02 g/mol
MDL Number: MFCD30489168
CAS Number: 1611002-56-2
UN: 3146
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR