4-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-1,3-dimethyl-7-azaindazole-5-carboxylic acid, 5-Carboxy-4-chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine

A168926-10G A168926-1G A168926-250MG
77286-730EA 306.94 USD
77286-730 77286-732 77286-734
4-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid 95%
4-Chloro-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈ClN₃O₂
MW: 225.632 g/mol
MDL Number: MFCD00067959
CAS Number: 175201-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR