Ky 05009 98+%

Supplier: Ambeed
Warning

Synonyms: KY-05009, 5-(4-Methylbenzamido)-2-(phenylamino)thiazole-4-carboxamide

A168785-10MG A168785-1MG A168785-25MG A168785-50MG A168785-5MG
77259-854EA 37.08 USD
77259-854 77259-856 77259-858 77259-860 77259-862
Ky 05009 98+%
Ky 05009

Precaución: for Research Use Only

Formula: C₁₈H₁₆N₄O₂S
MW: 352.41 g/mol
MDL Number: MFCD29059915
CAS Number: 1228280-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR