4-Amino-3-furazanecarboxamidoxime 97%
Supplier: Ambeed
|
Danger
|
A168482-10G
A168482-1G
A168482-250MG
A168482-25G
77286-700EA
157
USD
77286-700
77286-702
77286-704
77286-706
4-Amino-3-furazanecarboxamidoxime 97%
4-Amino-3-furazanecarboxamidoxime
Precaución: for Research Use Only
Formula:
C₃H₅N₅O₂ |
MDL Number:
MFCD00186407 CAS Number: 13490-32-9 UN: 1325 ADR: 4.1,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...