1-Butyl-3-(4-sulfobutyl)-1H-imidazol-3-ium trifluoromethanesulfonate 97%
Supplier: Ambeed
|
Warning
|
A168233-25G
A168233-5G
77247-918EA
64.89
USD
77247-918
77550-078
1-Butyl-3-(4-sulfobutyl)-1H-imidazol-3-ium trifluoromethanesulfonate 97%
1-Butyl-3-(4-sulfobutyl)-1H-imidazol-3-ium trifluoromethanesulfonate
Precaución: for Research Use Only
Formula:
C₁₂H₂₁F₃N₂O₆S₂ MW: 410.43 g/mol |
MDL Number:
MFCD07784446 CAS Number: 439937-63-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...