4-Fluoro-3'-iododiphenyl ether 95%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Fluorophenoxy)iodobenzene

A168215-1G A168215-5G
77286-684EA 57.68 USD
77286-684 77286-686
4-Fluoro-3'-iododiphenyl ether 95%
4-Fluoro-3'-iododiphenyl ether

Precaución: for Research Use Only

Formula: C₁₂H₈FIO
MW: 314.1 g/mol
MDL Number: MFCD00236611
CAS Number: 188534-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR