2-Chloro-5-hydroxynicotinic acid 95%

Supplier: Ambeed
Warning

A168032-100MG A168032-1G A168032-250MG A168032-5G
77259-826EA 80.34 USD
77259-826 77259-828 77259-830 77748-118
2-Chloro-5-hydroxynicotinic acid 95%
2-Chloro-5-hydroxynicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄ClNO₃
MW: 173.55 g/mol
MDL Number: MFCD09757509
CAS Number: 42959-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR