4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) 95%

Supplier: Ambeed
Warning

A168020-100G A168020-10G A168020-1G A168020-250MG A168020-25G A168020-5G
77286-672EA 267 USD
77286-672 77286-674 77286-676 77286-678 77286-680 77286-682
4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture) 95%
4-Hydroxycyclohexanecarboxylic acid (cis and trans mixture)

Precaución: for Research Use Only

Formula: C₇H₁₂O₃
MDL Number: MFCD00077974
CAS Number: 17419-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR