1-(4-(Benzyloxy)phenyl)-2-bromoethanone 98%

Supplier: Ambeed
Danger

A168007-100MG A168007-1G A168007-250MG
77286-666EA 17.51 USD
77286-666 77286-668 77286-670
1-(4-(Benzyloxy)phenyl)-2-bromoethanone 98%
1-(4-(Benzyloxy)phenyl)-2-bromoethanone

Precaución: for Research Use Only

Formula: C₁₅H₁₃BrO₂
MDL Number: MFCD07367994
CAS Number: 4254-67-5
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR