PEG5-carboxylic acid tert-butyl ester 97%

Supplier: Ambeed
Warning

Synonyms: HO-PEG4 tert-Butyl Ester, Hydroxy-PEG4-(CH2)2-Boc, tert-Butyl 1-Hydroxy-3,6,9,12-tetraoxapentadecan-15-oate, 1-Hydroxy-3,6,9,12-tetraoxapentadecan-15-oic acid tert-Butyl Ester

A167912-1G A167912-250MG A167912-25G A167912-5G
77247-896EA 24.72 USD
77247-896 77247-898 77247-900 77247-902
PEG5-carboxylic acid tert-butyl ester 97%
PEG5-carboxylic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₅H₃₀O₇
MW: 322.40 g/mol
MDL Number: MFCD11041117
CAS Number: 518044-32-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR