trans-4'-(3,4-Difluorophenyl)bicyclohexyl-4-one 97%

Supplier: Ambeed
Warning

Synonyms: trans-4'-(3,4-Difluorophenyl)-[1,1'-bi(cyclohexan)]-4-one

A167853-100G A167853-1G
77236-178EA 88.58 USD
77236-178 77236-180
trans-4'-(3,4-Difluorophenyl)bicyclohexyl-4-one 97%
trans-4'-(3,4-Difluorophenyl)bicyclohexyl-4-one

Precaución: for Research Use Only

Formula: C₁₈H₂₂F₂O
MW: 292.37 g/mol
MDL Number: MFCD26143179
CAS Number: 147622-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR