2-(4-Pyridyl)benzimidazole 98%
Supplier: Ambeed
|
Warning
|
A167748-1G
A167748-25G
A167748-5G
77236-172EA
13
USD
77236-172
77236-174
77236-176
2-(4-Pyridyl)benzimidazole 98%
2-(4-Pyridyl)benzimidazole
Precaución: for Research Use Only
Formula:
C₁₂H₉N₃ |
MDL Number:
MFCD00453948 CAS Number: 2208-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...