Methyl-2,2-dimethoxypropionate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,2-dimethoxypropanoate

A167678-10G A167678-1G A167678-25G A167678-5G
77247-884EA 116.39 USD
77247-884 77247-886 77247-888 77247-890
Methyl-2,2-dimethoxypropionate 98%
Methyl-2,2-dimethoxypropionate

Precaución: for Research Use Only

Formula: C₆H₁₂O₄
MW: 148.1571 g/mol
MDL Number: MFCD00048066
CAS Number: 10076-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR