3-(Boc-amino)-3-(4-fluorophenyl)propionic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-((tert-Butoxycarbonyl)amino)-3-(4-fluorophenyl)propanoic acid, 3-[(tert-Butoxycarbonyl)amino]-3-(4-fluorophenyl)propanoic acid

A167573-10G A167573-5G
77273-582EA 268.83 USD
77273-582 77273-584
3-(Boc-amino)-3-(4-fluorophenyl)propionic acid 97%
3-(Boc-amino)-3-(4-fluorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO₄
MW: 283.30 g/mol
MDL Number: MFCD02090715
CAS Number: 284493-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR