2,4-Dichloro-6-morpholino-1,3,5-triazine 97%

Supplier: Ambeed
Warning

Synonyms: 4-(4,6-Dichloro-1,3,5-triazin-2-yl)morpholine, 2,4-Dichloro-6-(4-morpholinyl)-1,3,5-triazine

A167530-10G A167530-1G A167530-5G A167530-25G
77247-878EA 35.02 USD
77247-878 77247-880 77247-882 77593-024
2,4-Dichloro-6-morpholino-1,3,5-triazine 97%
2,4-Dichloro-6-morpholino-1,3,5-triazine

Precaución: for Research Use Only

Formula: C₇H₈Cl₂N₄O
MW: 235.0706 g/mol
MDL Number: MFCD00460178
CAS Number: 6601-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR