Hydroxyterephthalic acid 97%

Supplier: Ambeed
Warning

A167506-10G A167506-1G A167506-250MG A167506-25G A167506-5G A167506-100G
77259-808EA 34 USD
77259-808 77259-810 77259-812 77259-814 77259-816 77567-442
Hydroxyterephthalic acid 97%
Hydroxyterephthalic acid

Precaución: For research use only.

Formula: C₈H₆O₅
MDL Number: MFCD09835368
CAS Number: 636-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR