trans-3-(4-Pyridyl)acrylic acid 95%

Supplier: Ambeed
Warning

A167141-1G A167141-25G A167141-5G
77259-802EA 32.96 USD
77259-802 77259-804 77259-806
trans-3-(4-Pyridyl)acrylic acid 95%
trans-3-(4-Pyridyl)acrylic acid

Precaución: for Research Use Only

Formula: C₈H₇NO₂
MDL Number: MFCD00040746
CAS Number: 84228-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR