Phenyl (3-methoxy-1,2,4-thiadiazol-5-yl)carbamate 97%
Supplier: Ambeed
|
Warning
|
A166979-100MG
A166979-250MG
77247-874EA
30.9
USD
77247-874
77247-876
Phenyl (3-methoxy-1,2,4-thiadiazol-5-yl)carbamate 97%
Phenyl (3-methoxy-1,2,4-thiadiazol-5-yl)carbamate
Precaución: for Research Use Only
Formula:
C₁₀H₉N₃O₃S MW: 251.26 g/mol |
MDL Number:
MFCD30489554 CAS Number: 1445988-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...