4-Benzyl-2-hydroxymorpholin-3-one 97%

Supplier: Ambeed
Warning

A166952-10G A166952-25G A166952-5G
77286-632EA 12 USD
77286-632 77286-634 77286-636
4-Benzyl-2-hydroxymorpholin-3-one 97%
4-Benzyl-2-hydroxymorpholin-3-one

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO₃
MDL Number: MFCD07776435
CAS Number: 287930-73-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR