(3,4,5-Trifluorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

A166941-100G A166941-10G A166941-1G A166941-250MG A166941-25G A166941-5G
77236-142EA 147 USD
77236-142 77236-144 77236-146 77236-148 77236-150 77236-152
(3,4,5-Trifluorophenyl)acetic acid 97%
(3,4,5-Trifluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₂F₃)CH₂COOH
MDL Number: MFCD00083532
CAS Number: 209991-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR