4,4',4'',4'''-([2,2'-Bi(1,3-dithiolylidene)]-4,4',5,5'-tetrayl)tetrabenzaldehyde 97%
Supplier: Ambeed
|
Warning
|
A166791-1G
A166791-250MG
A166791-100MG
A166791-50MG
77259-794EA
1253
USD
77259-794
77615-388
77748-102
77748-103
4,4',4'',4'''-([2,2'-Bi(1,3-dithiolylidene)]-4,4',5,5'-tetrayl)tetrabenzaldehyde 97%
4,4',4'',4'''-([2,2'-Bi(1,3-dithiolylidene)]-4,4',5,5'-tetrayl)tetrabenzaldehyde
Precaución: for Research Use Only
Formula:
C₃₄H₂₀O₄S₄ MW: 620.78 g/mol |
CAS Number:
1639134-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...