2-Adamantylamine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 2-Aminoadamantane hydrochloride, Tricyclo[3.3.1.1(3,7)]dec-2-ylamine hydrochloride, 2-Adamantanamine hydrochloride, Adamantan-2-amine hydrochloride, 2-Aminotricyclo[3.3.1.1(3,7)]decane hydrochloride, 2-Adamantanammonium chloride

A166662-1G A166662-250MG A166662-25G
77247-870EA 11.33 USD
77247-870 77247-872 77567-424
2-Adamantylamine hydrochloride 97%
2-Adamantylamine hydrochloride

Precaución: For research use only.

Formula: C₁₀H₁₈ClN
MW: 187.71 g/mol
MDL Number: MFCD00074743
CAS Number: 10523-68-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR