6-Methoxysalicylaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-6-methoxybenzaldehyde

A166631-10G A166631-1G A166631-250MG A166631-25G
77247-860EA 199.82 USD
77247-860 77247-862 77247-864 77247-866
6-Methoxysalicylaldehyde 98%
6-Methoxysalicylaldehyde

Precaución: for Research Use Only

Formula: C₈H₈O₃
MW: 152.15 g/mol
MDL Number: MFCD00151830
CAS Number: 700-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR