Benzo[d]thiazol-4-ol 98%

Supplier: Ambeed
Warning

A166589-250MG A166589-25G A166589-5G
77286-624EA 29.87 USD
77286-624 77286-626 77286-628
Benzo[d]thiazol-4-ol 98%
Benzo[d]thiazol-4-ol

Precaución: for Research Use Only

Formula: C₇H₅NOS
MW: 151.19 g/mol
MDL Number: MFCD11046398
CAS Number: 7405-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR