(2,4-Dimethylphenyl)(2-nitrophenyl)sulfane 98%

Supplier: Ambeed
Warning

A166575-1G A166575-25G A166575-5G
77247-850EA 9.27 USD
77247-850 77247-852 77247-854
(2,4-Dimethylphenyl)(2-nitrophenyl)sulfane 98%
(2,4-Dimethylphenyl)(2-nitrophenyl)sulfane

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₂S
MW: 259.32 g/mol
MDL Number: MFCD27918698
CAS Number: 1610527-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR