2,2,6-Trimethyl-4H-1,3-dioxin-4-one 95%

Supplier: Ambeed
Warning

A166559-100G A166559-500G A166559-5G A166559-25G
77247-844EA 26 USD
77247-844 77247-846 77247-848 77320-596
2,2,6-Trimethyl-4H-1,3-dioxin-4-one 95%
2,2,6-Trimethyl-4H-1,3-dioxin-4-one

Precaución: for Research Use Only

Formula: C₇H₁₀O₃
MDL Number: MFCD00040468
CAS Number: 5394-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR