1-Methylbenzimidazole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Methyl-1H-benzimidazole

A166252-100G A166252-10G A166252-25G
77247-826EA 207.03 USD
77247-826 77247-828 77247-830 77247-832 77247-834
1-Methylbenzimidazole 98%
1-Methylbenzimidazole

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.16 g/mol
MDL Number: MFCD00192275
CAS Number: 1632-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR