3-(Benzo[d][1,3]dioxol-5-ylmethylene)-2-oxopyrrolidine-1-carbaldehyde 98%

Supplier: Ambeed
Warning

A166203-100MG A166203-10MG A166203-1G A166203-1MG A166203-250MG A166203-25MG A166203-50MG A166203-5MG
77236-118EA 194.67 USD
77236-118 77236-120 77236-122 77236-124 77236-126 77236-128 77236-130 77236-132
3-(Benzo[d][1,3]dioxol-5-ylmethylene)-2-oxopyrrolidine-1-carbaldehyde 98%
3-(Benzo[d][1,3]dioxol-5-ylmethylene)-2-oxopyrrolidine-1-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₄
MW: 245.23 g/mol
MDL Number: MFCD03453551
CAS Number: 218924-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR