5-Chloro-2,2'-bipyridine 97%

Supplier: Ambeed
Warning

A166146-100MG A166146-250MG
77247-820EA 22 USD
77247-820 77247-822
5-Chloro-2,2'-bipyridine 97%
5-Chloro-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₁₀H₇ClN₂
MW: 190.63 g/mol
MDL Number: MFCD11112154
CAS Number: 162612-08-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR