4,5,6,7-Tetrachloro-2-hydroxyisoindoline-1,3-dione 97%

Supplier: Ambeed
Danger

Synonyms: N-Hydroxytetrachlorophthalimide

A166141-1G A166141-250MG A166141-25G A166141-5G
77247-812EA 38.11 USD
77247-812 77247-814 77247-816 77247-818
4,5,6,7-Tetrachloro-2-hydroxyisoindoline-1,3-dione 97%
4,5,6,7-Tetrachloro-2-hydroxyisoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₈HCl₄NO₃
MW: 300.91 g/mol
MDL Number: MFCD29088000
CAS Number: 85342-65-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR