2-Chloro-6-methoxybenzothiazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-6-methoxy-1,3-benzothiazole, 2-Chloro-6-methoxybenzo[d]thiazole

A166136-10G A166136-1G A166136-25G A166136-5G A166136-250MG
77236-110EA 128.75 USD
77236-110 77236-112 77236-114 77236-116 77746-122
2-Chloro-6-methoxybenzothiazole 97%
2-Chloro-6-methoxybenzothiazole

Precaución: for Research Use Only

Formula: C₈H₆ClNOS
MW: 199.66 g/mol
MDL Number: MFCD00671510
CAS Number: 2605-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR