Methyl 2-methyl-1-oxo-1,2,3,4-tetrahydronaphthalene-2-carboxylate 98%

Supplier: Ambeed
Warning

A166094-1G A166094-250MG
77247-808EA 18 USD
77247-808 77247-810
Methyl 2-methyl-1-oxo-1,2,3,4-tetrahydronaphthalene-2-carboxylate 98%
Methyl 2-methyl-1-oxo-1,2,3,4-tetrahydronaphthalene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₄O₃
MDL Number: MFCD20233605
CAS Number: 54125-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR