5-Methoxy-2-nitrobenzaldehyde 98%

Supplier: Ambeed
Warning

A166000-10G A166000-1G A166000-250MG A166000-25G A166000-5G
77236-098EA 42 USD
77236-098 77236-100 77236-102 77236-104 77236-106
5-Methoxy-2-nitrobenzaldehyde 98%
5-Methoxy-2-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇NO₄
MDL Number: MFCD00456498
CAS Number: 20357-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR