N-(tert-Butoxycarbonyl)-D-threonine 95%

Supplier: Ambeed
Warning

Synonyms: N-Boc-D-threonine, Boc-D-Thr-OH, (2R,3S)-2-((tert-Butoxycarbonyl)amino)-3-hydroxybutanoic acid

A165896-10G A165896-1G A165896-250MG A165896-25G A165896-5G A165896-100G
77259-774EA 57.68 USD
77259-774 77259-776 77259-778 77259-780 77615-378 77746-118
N-(tert-Butoxycarbonyl)-D-threonine 95%
N-(tert-Butoxycarbonyl)-D-threonine

Precaución: for Research Use Only

Formula: C₉H₁₇NO₅
MW: 219.24 g/mol
MDL Number: MFCD00037807
CAS Number: 55674-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR