1-Ethyl-4-iodobenzene 96%

Supplier: Ambeed
Warning

Synonyms: 4-Ethyliodobenzene

A165816-100G A165816-1G A165816-25G A165816-5G
77236-080EA 141.11 USD
77236-080 77236-082 77236-084 77236-086
1-Ethyl-4-iodobenzene 96%
1-Ethyl-4-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₉I
MW: 232.06 g/mol
MDL Number: MFCD00060663
CAS Number: 25309-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR