2-Amino-4-methyl-3-nitropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-3-nitro-2-pyridylamine, 4-Methyl-3-nitropyridin-2-amine, 2-Amino-3-nitro-4-picoline

A165711-100G A165711-10G A165711-25G A165711-5G
77273-530EA 127 USD
77273-530 77273-532 77273-534 77273-536
2-Amino-4-methyl-3-nitropyridine 98%
2-Amino-4-methyl-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Melting Pt: 140
MDL Number: MFCD00006315
CAS Number: 6635-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR