2,4-Dimethylbenzyl alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (2,4-Dimethylphenyl)methanol, 4-(Hydroxymethyl)-m-xylene

A165671-1G A165671-25G A165671-5G
77247-802EA 18.54 USD
77247-802 77247-804 77247-806
2,4-Dimethylbenzyl alcohol 97%
2,4-Dimethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₂O
MW: 136.19 g/mol
MDL Number: MFCD00004624
CAS Number: 16308-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR