2,4-Dimethylbenzoyl chloride 97%

Supplier: Ambeed
Danger

A165541-250MG A165541-5G
77247-796EA 18 USD
77247-796 77247-798
2,4-Dimethylbenzoyl chloride 97%
2,4-Dimethylbenzoyl chloride

Precaución: for Research Use Only

Formula: C₉H₉ClO
MDL Number: MFCD00798005
CAS Number: 21900-42-5
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR