2-Fluoro-1-iodo-4-nitrobenzene 95%

Supplier: Ambeed
Warning

A165510-100MG A165510-10G A165510-1G A165510-250MG A165510-25G A165510-5G
77286-596EA 9.27 USD
77286-596 77286-598 77286-600 77286-602 77286-604 77286-606
2-Fluoro-1-iodo-4-nitrobenzene 95%
2-Fluoro-1-iodo-4-nitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃FINO₂
MW: 267 g/mol
MDL Number: MFCD00033987
CAS Number: 2996-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR