Methyl-4-aminosalicylate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-amino-2-hydroxybenzoate

A165416-10G A165416-25G A165416-5G A165416-100G
77247-790EA 8.24 USD
77247-790 77247-792 77247-794 77746-116
Methyl-4-aminosalicylate 97%
Methyl 4-aminosalicylate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.18 g/mol
MDL Number: MFCD00088091
CAS Number: 4136-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR