2-Phenylthiazole 95%

Supplier: Ambeed
Warning

A165300-1G A165300-25G A165300-5G
77273-506EA 33.99 USD
77273-506 77273-508 77273-510
2-Phenylthiazole 95%
2-Phenylthiazole

Precaución: for Research Use Only

Formula: C₉H₇NS
MW: 161.2200 g/mol
MDL Number: MFCD00272327
CAS Number: 1826-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR