1-Benzyloxy-4-iodobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Benzyloxy)-4-iodobenzene, 4-Benzyloxyiodobenzene

A165244-10G A165244-1G A165244-25G A165244-5G
77286-588EA 95.79 USD
77286-588 77286-590 77286-592 77286-594
1-Benzyloxy-4-iodobenzene 98%
1-Benzyloxy-4-iodobenzene

Precaución: for Research Use Only

Formula: C₁₃H₁₁IO
MW: 310.13 g/mol
MDL Number: MFCD00079704
CAS Number: 19578-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR