7-Bromo-2,2-dimethylchroman-4-one 97%

Supplier: Ambeed
Warning

A165204-1G A165204-250MG
77247-778EA 115.36 USD
77247-778 77247-780
7-Bromo-2,2-dimethylchroman-4-one 97%
7-Bromo-2,2-dimethylchroman-4-one

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO₂
MW: 255.10 g/mol
MDL Number: MFCD13190381
CAS Number: 130200-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR