4-Bromo-9,9'-spirobi[9H-fluorene] 98%

Supplier: Ambeed
Warning

A165157-10G A165157-1G A165157-25G A165157-5G
77236-056EA 36 USD
77236-056 77236-058 77236-060 77236-062
4-Bromo-9,9'-spirobi[9H-fluorene] 98%
4-Bromo-9,9'-spirobi[9H-fluorene]

Precaución: for Research Use Only

Formula: C₂₅H₁₅Br
MDL Number: MFCD16660920
CAS Number: 1161009-88-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR