3-Bromo-9,9-diphenyl-9H-fluorene 98%

Supplier: Ambeed
Warning

A165017-100MG A165017-1G A165017-250MG A165017-5G
77236-048EA 6.18 USD
77236-048 77236-050 77236-052 77236-054
3-Bromo-9,9-diphenyl-9H-fluorene 98%
3-Bromo-9,9-diphenyl-9H-fluorene

Precaución: for Research Use Only

Formula: C₂₅H₁₇Br
MW: 397.32 g/mol
MDL Number: MFCD24849694
CAS Number: 1547491-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR